Skip to Content
Merck
All Photos(1)

Key Documents

111457

Sigma-Aldrich

2,2-Dimethylhexane

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3C(CH3)3
CAS Number:
Molecular Weight:
114.23
Beilstein:
1696861
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

1.22 psi ( 37.7 °C)

Quality Level

Assay

98%

refractive index

n20/D 1.393 (lit.)

bp

107 °C (lit.)

density

0.693 g/mL at 25 °C (lit.)

SMILES string

CCCCC(C)(C)C

InChI

1S/C8H18/c1-5-6-7-8(2,3)4/h5-7H2,1-4H3

InChI key

FLTJDUOFAQWHDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,2-Dimethylhexane is one of the products of the corona discharge of a simulated Titan′s atmosphere (2% argon in nitrogen and 10% methane) at 500 Torr and 298 K (using GC-FTIR-MS techniques). It was used to study the adsorption of linear and branched alkanes on a microporous silicoaluminophosphate material (SAPO-5) using chromatographic techniques.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

26.6 °F - closed cup

Flash Point(C)

-3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Navarro-Gonzalez et al.
Advances in space research : the official journal of the Committee on Space Research (COSPAR), 19(7), 1121-1133 (1997-01-01)
The atmosphere of Titan is constantly bombarded by galactic cosmic rays and Saturnian magnetospheric electrons causing the formation of free electrons and primary ions, which are then stabilized by ion cluster formation and charging of aerosols. These charged particles accumulate
Todd C Atwood et al.
Scientific reports, 7(1), 13193-13193 (2017-10-19)
Recent decline of sea ice habitat has coincided with increased use of land by polar bears (Ursus maritimus) from the southern Beaufort Sea (SB), which may alter the risks of exposure to pathogens and contaminants. We assayed blood samples from
Fangyuan Zhou et al.
The ISME journal, 11(12), 2809-2820 (2017-08-12)
Interactions among microbial symbionts have multiple roles in the maintenance of insect-microbe symbiosis. However, signals mediating microbial interactions have been scarcely studied. In the classical model system of bark beetles and fungal associates, fungi increase the fitness of insects. However
Kang Jian et al.
Environmental pollution (Barking, Essex : 1987), 258, 113737-113737 (2019-12-16)
Food products are inevitably contaminated by flame retardants throughout their lifecycle (i.e., during production, use, and disposal). In order to evaluate the dietary intake of legacy and emerging halogenated flame retardants (HFRs) in typical market food in China, we investigate
Xue Xiong et al.
Journal of chromatography. A, 1567, 191-197 (2018-08-14)
This work describes the separation performance of utilizing hexagonal boron nitride (h-BN) as the stationary phase for capillary gas chromatography (GC). The statically coated h-BN column showed moderate polarity and achieved column efficiencies of 3455 plates/m and 3800 plates/m for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service