Skip to Content
Merck
All Photos(2)

Key Documents

109967

Sigma-Aldrich

2-Ethoxyethyl acetate

98%

Synonym(s):

1-Acetoxy-2-ethoxyethane, Cellosolve® acetate, Ethylene glycol monoethyl ether acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH2CH2OC2H5
CAS Number:
Molecular Weight:
132.16
Beilstein:
1748677
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
98%
technique(s):
microbiological culture: suitable
bp:
156 °C (lit.)
vapor pressure:
2 mmHg ( 20 °C)

vapor density

4.6 (vs air)

Quality Level

vapor pressure

2 mmHg ( 20 °C)

Assay

98%

form

liquid

autoignition temp.

715 °F

expl. lim.

13 %

technique(s)

microbiological culture: suitable

refractive index

n20/D 1.403-1.409
n20/D 1.406 (lit.)

bp

156 °C (lit.)

mp

−61 °C (lit.)

solubility

H2O: soluble about 6 parts

density

0.975 g/mL at 25 °C (lit.)

functional group

ester
ether

SMILES string

CCOCCOC(C)=O

InChI

1S/C6H12O3/c1-3-8-4-5-9-6(2)7/h3-5H2,1-2H3

InChI key

SVONRAPFKPVNKG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Ethoxyethyl acetate is used to study the field evaluation of a passive badge for measuring 2-ethoxyethyl acetate.

Legal Information

Cellosolve is a registered trademark of Union Carbide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

S E Chia et al.
American journal of industrial medicine, 31(2), 148-152 (1997-02-01)
The objectives of this study were to determine the exposure levels among workers who handle 2-ethoxyethylacetate (EGEEA) in the liquid crystal display (LCD) manufacturing industry and to study the menstrual patterns among the exposed workers compared to a referent group
Y Kim et al.
Occupational and environmental medicine, 56(6), 378-382 (1999-09-04)
To evaluate exposure to mixed solvents containing ethylene glycol monoethyl ether acetate (EGEEA) in shipyard painters, to determine if EGEEA is toxic to the bone marrow. An industrial hygiene survey was performed to identify exposure to EGEEA of two groups
Reproductive toxicity of ethylene glycol monomethyl ether, ethylene glycol monoethyl ether and their acetates.
J A Wess
Scandinavian journal of work, environment & health, 18 Suppl 2, 43-45 (1992-01-01)
L M Sweeney et al.
Toxicological sciences : an official journal of the Society of Toxicology, 62(1), 124-139 (2001-06-16)
Methoxyethanol (ethylene glycol monomethyl ether, EGME), ethoxyethanol (ethylene glycol monoethyl ether, EGEE), and ethoxyethyl acetate (ethylene glycol monoethyl ether acetate, EGEEA) are all developmental toxicants in laboratory animals. Due to the imprecise nature of the exposure data in epidemiology studies
J E Doe
Environmental health perspectives, 57, 33-41 (1984-08-01)
An inhalation teratology study was conducted in rats at 10, 50 and 250 ppm ethylene glycol monoethyl ether (EGEE) and in rabbits at 10, 50 and 175 ppm EGEE. This study was designed to supplement a study conducted for NIOSH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service