Terpenes in Cannabis: Profile and Importance
![Terpenes testing in cannabis and hemp Profiling terpenes in cannabis to determine the purity, quality and identity of cannabis strains by using GC-MS while meeting legal and regulatory requirements.](/deepweb/assets/sigmaaldrich/marketing/global/images/applications/environmental-testing-and-industrial-hygiene/terpenes-testing-in-cannabis-and-hemp/terpenes-testing-in-cannabis-and-hemp.jpg)
Terpene profiling is fundamental to showing the identity and medical efficacy of cannabis cultivars or strains. Terpenes are the organic primary components of cannabis resins and essential oils with characteristic odor and distinctive organoleptic properties. Made up of volatile and semivolatile combinations of C5 isoprene subunits, terpenes are proposed to synergistically boost the therapeutic effects of cannabinoids by “entourage effect”. As the type and amounts of terpenes differ among genetic lineages of cannabis such as Cannabis sativa, Cannabis indica and Cannabis ruderalis, robust analytical methods and standards are in demand for identification of cultivars, and scientific validation of their medicinal and pharmacological properties.
Featured Categories
Browse our Gas Chromatography (GC) units, columns, consumables, and accessories for your analytical GC applications to achieve high performance, maximum productivity, and superior detection sensitivity.
Solid phase microextraction (SPME) is a solvent-free sample preparation technique. View our complete offering of SPME products. Our innovative products for SPME include the smart SPME fibers, nitinol-core fibers, and BioSPME.
Analytical standards and certified reference materials for analysis of terpenes, cannabinoids, pesticides, mycotoxins, heavy metals, and cannabis potency testing.
We offer a wide selection of autosampler and general-purpose vials, including septa, caps, accessories, amber and clear bottles, headspace vials, polypropylene and TPX vials, center drain vials, and silane-treated vials for special applications.
LEGAL AND REGULATORY REQUIREMENTS FOR TERPENES TESTING IN CANNABIS
The increased use of cannabis as a recreational or medicinal drug calls for stringent regulations for manufacturing practices and quality standards derived from cannabis source materials. To address concerns over the therapeutic efficacy, safety and quality of cannabis products, cannabis quality control testing must be carried out for terpenes across geographies, including the United States and Canada, where the landscape of legalization is rapidly changing.
WORKFLOW FOR PROFILING OF CANNABIS TERPENES
The typical workflow for terpene analysis in cannabis, hemp and their products includes sample preparation by solvent extraction or headspace-based solid-phase microextraction, preparation of standards and calibration,, gas chromatographic separation followed by mass spectrometric detection and data analysis of the terpene profile.
CANNABIS TERPENES SAMPLE PREPARATION BY EXTRACTION OR HEADSPACE-SPME
Testing laboratories today prominently use one of the following two techniques for sample preparation in analysis of cannabis terpenes: Simplified solvent extraction or headspace-based solid-phase microextraction (HS-SPME) . Sample preparation typically precedes GC-FID or GC-MS analysis.
HS-SPME technique offers the following benefits:
- ensuring nonvolatile matrix components are not co-extracted
- providing results with increased sensitivity
- simultaneously determining cannabis terpenes and residual solvents, remaining after their use in extraction with alcohols and hydrocarbons
- eliminating the need for special instrumentation
- amenable either manual or autosampler based sample preparation
Solvent extraction also doesn’t require special instrumentation and is a convenient alternative when matrix interferences are not critical.
CALIBRATION STANDARDS FOR CANNABIS TERPENE QUANTITATION
Profiling and quantitation of terpenes in cannabis products requires the use of authentic and accurate reference standards. Given the isomeric properties of many terpenes, references that characterize the isomer or enantiomer may be important for identification and quantitation. Certified reference materials (CRMs) manufactured according to ISO17034 offer accuracy of concentration with uncertainty and traceability and are recommended for accurate quantitation. CRMs are typically diluted into working standards and calibration curves in the reference range for analysis of the sample.
GC-MS ANALYSIS OF TERPENES IN CANNABIS
GC-MS and GC-FID based methods are at the forefront of terpene profiling and quantitation in cannabis terpene testing with excellent ionization response under electron ionization (EI) sources.
GC-MS is an efficient and robust technique to investigate cannabis products for terpenes, providing chromatographic resolution, identification and quantitation. Chiral GC columns can enhance identification by overcoming the challenges posed by isomers and differences in the aromatic character of terpenes. GC x GC techniques are suitable for terpene profiling in complex cannabis matrices, with retention time indexing for greater accuracy.
The use of high-quality standards, reagents and chromatography consumables is a prerequisite for accurate and reliable testing and detection of terpenes in cannabis crops and cannabis-derived products. The correct choice of CRMs, analytical reference standards, high-purity solvents as well as columns form the basis of method development and validation, instrument calibration, quality control and system suitability check.
Visit our document search for data sheets, certificates and technical documentation.
Related Articles
- Reference materials, standards and certified reference materials are critical for analytical testing. We can help you to choose the ideal reference material for your application, whether you are using reference materials for calibration or quantitative analysis.
- ISO-certified cannabinoid CRMs, THC and THCA, available as single components or mixes, for accurate potency testing.
- A step-by-step workflow for the sample preparation of hemp bud, hemp oil, chocolate, gummy, candy, and cream for the HPLC-PDA separation and analysis of 17 cannabinoids.
- A 4-step comprehensive ICP-MS workflow including sample homogenization, with standard addition calibration method, to determine heavy metals in Cannabis sativa.
- Learn more about how the thermal desorption technique is applied for terpene identification, profiling, odor measurement, industrial hygiene, and environmental emissions.
- See All (4)
Related Protocols
- Complete workflow for the comprehensive analysis of terpenes in cannabis
- SPME offers efficient sample preparation for analyzing terpene/terpenoid profiles in plant essential oils using automation.
- GC Analysis of Terpenes in Cannabis on Equity®-1 after Headspace SPME using 50/30 μm DVB/CAR/PDMS Fiber
- Karl Fischer titration offers accurate water content determination in hemp samples compared to the loss on drying method.
- Simplify your thermal desorption-based terpenes sample collection in air. Learn more about their profiling, odor measurement, industrial hygiene, and environmental emissions.
- See All (5)
Find More Articles and Protocols
How Can We Help
In case of any questions, please submit a customer support request
or talk to our customer service team:
Email [email protected]
or call +1 (800) 244-1173
Additional Support
- Calculators & Apps
Web Toolbox - science research tools and resources for analytical chemistry, life science, chemical synthesis and materials science.
- Customer Support Request
Customer support including help with orders, products, accounts, and website technical issues.
- FAQ