Merck
Protein Molecular Weight Calibration Curve on Zenix® Size Exclusion Columns, Effect of Pore Size
Protein Molecular Weight Calibration Curve on Zenix® Size Exclusion Columns, Effect of Pore Size application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Zenix® SEC-150 Gel Filtration Column, 3 μm
L × I.D. 30 cm × 7.8 mm, 3 μm particle size, stainless steel
Zenix® SEC-300 Gel Filtration Column, 3 μm
L × I.D. 30 cm × 7.8 mm, 3 μm particle size, stainless steel
standard
Product No.
Description
Pricing
γ-Globulins from human blood
≥99% (electrophoresis)
Myoglobin from equine heart
≥90% (SDS-PAGE), essentially salt-free, lyophilized powder
Vitamin B12
≥98%
Thyroglobulin from bovine thyroid
powder, ≥90% (agarose gel electrophoresis)
Bovine Serum Albumin
lyophilized powder, ≥96% (agarose gel electrophoresis)
Ribonuclease A from bovine pancreas
Molecular Biology, ≥70 Kunitz units/mg protein, lyophilized
Ovalbumin (257-264) chicken
≥97% (HPLC)
CONDITIONS
column
Zenix SEC-150, 30 cm x 7.8 mm I.D., 3 μm, 150 Å particles (Z777018)
column
Zenix SEC-300, 30 cm x 7.8 mm I.D., 3 μm, 300 Å particles (Z777033)
mobile phase
150 mM sodium phosphate, pH 7
flow rate
1 mL/min
column temp.
ambient
detector
UV, 214 nm
injection
10 μL
sample
each 1 mg/mL except uracil, 2.5 mg/mL
Description
Legal Information
Zenix is a registered trademark of Sepax Technologies