LC/MS Analysis of Cyanuric Acid on Ascentis® Express HILIC
Materials
analytical column
Product No.
Description
Pricing
mobile phase component
Product No.
Description
Pricing
CONDITIONS
column
Ascentis Express HILIC, 5 cm x 2.1 mm I.D., 2.7 μm particles (53934-U)
mobile phase
5 mM ammonium formate in 95:5 (v/v) acetonitrile:water
flow rate
0.2 mL/min
pressure
328 psi (22.6 bar)
column temp.
35 °C
detector
MS, ESI(-), SIR mode
injection
2 μL
sample
1 μg/mL in mobile phase
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany