Merck
HPLC Analysis of Tröger′s Base Enantiomers on Astec® Cellulose DMP (LC/MS Mobile Phase)
HPLC Analysis of Tröger′s Base Enantiomers on Astec® Cellulose DMP (LC/MS Mobile Phase) application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® Cellulose DMP Chiral HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
standard
Product No.
Description
Pricing
Troger′s Base
98%
CONDITIONS
column
Astec Cellulose DMP, 15 cm x 4.6 mm I.D., 5 μm particles (51098AST)
mobile phase
[A] methanol; [B] ammonium formate; (100:0.1, A:B)
flow rate
0.5 mL/min
column temp.
25 °C
detector
UV, 230 nm
injection
2 μL
Description
Application
Tröger′s base is an organic compound that exhibits chirality due to the presence of two bridgehead stereogenic nitrogen atoms in its structure.