HPLC Analysis of Hemoglobin Variants on Discovery® BIO PolyMA-WAX
![HPLC Analysis of Hemoglobin Variants on Discovery® BIO PolyMA-WAX application for HPLC](/deepweb/assets/sigmaaldrich/product/images/253/584/63bf8c66-88cc-4dac-bf47-f28b9c99b137/800/63bf8c66-88cc-4dac-bf47-f28b9c99b137.jpg)
Materials
standard
Product No.
Description
Pricing
CONDITIONS
column
Discovery BIO PolyMA-WAX, 5 cm x 4.6 mm, 5 μm particles (59602-U)
mobile phase
[A] 10 mM Tris, pH 8.0 with acetic acid; [B] 10 mM Tris, 0.25 M KCl, pH 8.0 with acetic acid
gradient
0 to 32% B in 20 min. (linear)
flow rate
0.5 mL/min
column temp.
35 °C
detector
UV, 280 nm
injection
50 μg each Hb variants (A2, S and A0)
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany