HPLC Analysis of Cysteine Enantiomers on Astec® CHIROBIOTIC® T (Formic Acid Mobile Phase Additive)
Materials
analytical column
Product No.
Description
Pricing
guard cartridge
Product No.
Description
Pricing
Astec® CHIROBIOTIC® T Chiral HPLC Guard Column
Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mmstandard
Product No.
Description
Pricing
L-Cysteine
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, SwitzerlandCONDITIONS
column
Astec CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 μm particles (12024AST)
mobile phase
[A] water; [B] methanol; [C] formic acid; (40:60:0.02, A:B:C)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 205 nm
injection
10 μL
sample
(200 μg /mL in 50:50 water:methanol)
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC