HPLC Analysis of Benzoic Acid and Sorbic Acid on Ascentis® Express RP-Amide ("Green" Mobile Phase Conditions)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Ascentis Express RP-Amide, 5 cm x 3 mm I.D., 2.7 μm particles (53916-U)
mobile phase
[A] 10 mM ammonium phosphate monobasic, pH 2.5 with phosphoric acid; [B] ethanol; (75:25, A:B)
flow rate
0.4 mL/min
pressure
138 bar (2000 psi) column, 248 bar (3600 psi) total
column temp.
55 °C
detector
UV, 215 nm
injection
1 μL
sample
50 μg/mL in mobile phase
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany