Merck
  • Levels of metamidophos in air and vegetables after greenhouse applications by gas chromatography.
Levels of metamidophos in air and vegetables after greenhouse applications by gas chromatography.
Journal of chromatography. A (1999-01-29)
F J Egea González, J L Martínez Vidal, M L Castro Cano, M Martínez Galera
ABSTRACT
The diminution of metamidophos residue levels with time in vegetables an greenhouse air was investigated after treatment of tomatoes and green beans. A gas chromatographic method using dichloromethane as an extraction solvent has been developed to analyse metamidophos in vegetables, with obtained recoveries higher than 89%. The reliability of several sorbents for air sampling was tested using standard atmospheres resulting in recoveries higher than 90% from PUF, XAD-2, XAD-4 and Supelpak using Soxhlet extraction with acetone. The dissipation of metamidophos in greenhouse air was studied 52 h after application. Finally the effect of crop, type of greenhouse, season and dose applied on the dissipation kinetic of metamidophos in vegetables, was statisTically studied by analysis of variance resulting in crop and season being the most significant factors.
Product Description
Supelco
ORBO 23 2-HMP on Amberlite XAD®-2 (20/40), 120/60 mg, W,W,W separators, O.D. × L 6 mm × 85 mm, pkg of 25 ea
Supelco
ORBO 49P (OVS) Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 270/140 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS) Tube, pkg of 10 ea
Supelco
Supelpak-2SVM, pkg of 500 g
Supelco
Supelpak-2, matrix styrene-divinylbenzene, 20-60 mesh, jar of 1000 g
Supelco
ORBO 43 Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 100/50 mg, W,W,W (specially treated) separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
Supelpak-2, matrix styrene-divinylbenzene, 20-60 mesh, jar of 100 g
Supelco
ORBO 24 2-HMP on Amberlite XAD®-2 (20/40), 150/75 mg, W,W,W separators, O.D. × L 6 mm × 105 mm, pkg of 25 ea