Merck
  • Diminished expression of CRHR2 in human colon cancer promotes tumor growth and EMT via persistent IL-6/Stat3 signaling.
Diminished expression of CRHR2 in human colon cancer promotes tumor growth and EMT via persistent IL-6/Stat3 signaling.
Cellular and molecular gastroenterology and hepatology (2015-10-27)
Jorge A Rodriguez, Sara Huerta-Yepez, Ivy Ka Man Law, Guillermina J Baay-Guzman, Belen Tirado-Rodriguez, Jill M Hoffman, Dimitrios Iliopoulos, Daniel W Hommes, Hein W Verspaget, Lin Chang, Charalabos Pothoulakis, Stavroula Baritaki
ABSTRACT
Chronic inflammation promotes development and progression of colorectal cancer (CRC). We explored the distribution of Corticotropin-Releasing-Hormone (CRH)-family of receptors and ligands in CRC and their contribution in tumor growth and oncogenic EMT. mRNA expression of CRH-family members was analyzed in CRC (N=56) and control (N=46) samples, 7 CRC cell lines and normal NCM460 cells. Immunohistochemical detection of CRHR2 was performed in 20 CRC and 5 normal tissues. Cell proliferation, migration and invasion were compared between Urocortin-2 (Ucn2)-stimulated parental and CRHR2-overexpressing (CRHR2+) cells in absence or presence of IL-6. CRHR2/Ucn2-targeted effects on tumor growth and EMT were validated in SW620-xenograft mouse models. CRC tissues and cell lines showed decreased mRNA and protein CRHR2 expression compared to controls and NCM460, respectively. The opposite trend was shown for Ucn2. CRHR2/Ucn2 signaling inhibited cell proliferation, migration, invasion and colony formation in CRC-CRHR2+ cells. In vivo, SW620-CRHR2+ xenografts showed decreased growth, reduced expression of EMT-inducers and elevated levels of EMT-suppressors. IL-1b, IL-6 and IL-6R mRNAs where diminished in CRC-CRHR2+ cells, while CRHR2/Ucn2 signaling inhibited IL-6-mediated Stat3 activation, invasion, migration and expression of downstream targets acting as cell cycle- and EMT-inducers. Expression of cell cycle- and EMT-suppressors was augmented in IL-6/Ucn2-stimulated CRHR2+ cells. In patients, CRHR2 mRNA expression was inversely correlated with IL-6R and vimentin levels and metastasis occurrence, while positively associated with E-cadherin expression and overall survival. CRHR2 downregulation in CRC supports tumor expansion and spread through maintaining persistent inflammation and constitutive Stat3 activation. CRHR2low CRC phenotypes are associated with higher risk for distant metastases and poor clinical outcomes.
Product Description
Roche
Cell Proliferation Kit II (XTT), liquid, pkg of 1 kit, suitable for cell analysis, suitable for tissue culture
Roche
In Situ Cell Death Detection Kit, POD, sufficient for ≤50 tests
Sigma-Aldrich
Anti-ZEB1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Corticotropin-Releasing Factor Receptor 2 Antibody, 1.0 mg/mL, from rabbit