Merck
  • Helicobacter pylori CagL dependent induction of gastrin expression via a novel αvβ5-integrin-integrin linked kinase signalling complex.
Helicobacter pylori CagL dependent induction of gastrin expression via a novel αvβ5-integrin-integrin linked kinase signalling complex.
Gut (2012-01-31)
Tobias Wiedemann, Stefan Hofbaur, Nicole Tegtmeyer, Sylwia Huber, Norbert Sewald, Silja Wessler, Steffen Backert, Gabriele Rieder
ABSTRACT
One of the most important hormones in the human stomach is the peptide gastrin. It is mainly required for the regulation of gastric pH but is also involved in growth and differentiation of gastric epithelial cells. In Helicobacter pylori infected patients, gastrin secretion can be upregulated by the pathogen, resulting in hypergastrinaemia. H pylori induced hypergastrinaemia is described as being a major risk factor for the development of gastric adenocarcinoma. In this study, the upstream receptor complex and bacterial factors involved in H pylori induced gastrin gene expression were investigated, utilising gastric epithelial cells which were stably transfected with a human gastrin promoter luciferase reporter construct. Integrin linked kinase (ILK) and integrin β5, but not integrin β1, played an important role in gastrin promoter activation. Interestingly, a novel CagL/integrin β5/ILK signalling complex was characterised as being important for H pylori induced gastrin expression. On interaction of H pylori with αvβ(5)-integrin and ILK, the epidermal growth factor receptor (EGFR)→Raf→mitogen activated protein kinase kinase (MEK)→extracellular signal regulated kinase (Erk) downstream signalling cascade was identified which plays a central role in H pylori gastrin induction. The newly discovered recognition receptor complex could be a useful target in treating precancerous conditions triggered by H pylori induced hypergastrinaemia.
Sigma-Aldrich
GeneJuice® Transfection Reagent, Non-lipid based chemical transfection reagent optimized for maximum transfection efficiency, ease-of-use, and minimal cytotoxicity on a wide variety of mammalian cells.
Sigma-Aldrich
Anti-Integrin αVβ5 Antibody, clone P1F6, azide free, clone P1F6, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-Interleukin-1β antibody produced in mouse, clone 8516.311, purified immunoglobulin, lyophilized powder
Sigma-Aldrich
Anti-Integrin β3 Antibody, clone B3A, azide free, clone B3A, Chemicon®, from mouse