Merck
  • Silica stationary phase functionalized by 4-carboxy-benzoboroxole with enhanced boronate affinity nature for selective capture and separation of cis-diol compounds.
Silica stationary phase functionalized by 4-carboxy-benzoboroxole with enhanced boronate affinity nature for selective capture and separation of cis-diol compounds.
Analytica chimica acta (2017-09-03)
Hengye Li, Xuemeng Zhang, Lin Zhang, Weihua Cheng, Fenying Kong, Dahe Fan, Lei Li, Wei Wang
ABSTRACT
4-Carboxy-benzoboroxole was designed and synthesized. It was then combined with the modification effect of polyethyleneimine (PEI) for the preparation of boronate affinity silica stationary phase. The stationary phase showed improved binding strength with dissociation constant (K
Sigma-Aldrich
Methyl 4-bromo-3-methylbenzoate, 95%
Sigma-Aldrich
HBTU, ≥98.0% (T)
Sigma-Aldrich
1-Hydroxy-7-azabenzotriazole solution, ~0.6 M in DMF