Merck
  • Bioanalytical LC-MS/MS method validation for plasma determination of topiramate in healthy Indian volunteers.
Bioanalytical LC-MS/MS method validation for plasma determination of topiramate in healthy Indian volunteers.
Biomedical chromatography : BMC (2009-07-14)
Dipanjan Goswami, Ajay Kumar, Arshad H Khuroo, Tausif Monif, Shamsur Rab
ABSTRACT
A LC-MS/MS method for plasma topiramate analysis is delineated involving least number of healthy volunteers. Topiramate and amlodipine internal standard (IS) were extracted by simple centrifuge-coupled solid-phase extraction and reverse-phase chromatographic separation was performed on an Ascentis C(18) column. Turbo-spray negative-ion mode multiple-reaction monitoring was selected for mass pair detection at m/z 338.3 --> 78.0 and m/z 407.3 --> 295.5 for analyte and IS respectively. The method showed a dynamic linearity range from 10.4 to 2045.0 ng/mL, lower limit of quantitation achieved at 10.4 ng/mL and finally a mass spectrometric total run time of within 2.5 min for human sample analysis. Bioequivalence was assessed successfully using this fully validated method on 16 fasted Indian male subjects with 25 mg topiramate tablet administration. An appropriate study design describes plasma samples collection up to 216 h post dose in two periods, separated by a 28 day washout period. The challenge of half-life matching for test and reference drug was achieved with 73.43 +/- 9.68 and 73.06 +/- 14.03 h, respectively, and intra-subject coefficient of variation achieved within 11% for AUCs and C(max) evaluated by non-compartmental pharmacokinetic analysis. The results of LCMS topiramate complete method validation supported by pharmacokinetic study have not been published before, and are presented and discussed for the first time in this article.
Product Description
Supelco
Ascentis® C18 HPLC Column, 3 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Ascentis® C18 HPLC Column, 3 μm particle size, L × I.D. 10 cm × 3 mm
Supelco
Ascentis® C18 HPLC Column, 3 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Ascentis® C18 HPLC Column, 3 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Ascentis® C18 HPLC Column, 3 μm particle size, L × I.D. 10 cm × 4.6 mm
Supelco
Ascentis® C18 HPLC Column, 3 μm particle size, L × I.D. 5 cm × 2.1 mm
Supelco
Ascentis® C18 HPLC Column, 3 μm particle size, L × I.D. 5 cm × 4.6 mm
Supelco
Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 21.2 mm
Supelco
Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 10 mm
Supelco
Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm
Supelco
Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 21.2 mm
Supelco
Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Ascentis® C18 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 4.6 mm