Merck
Phosphatases at the heart of FoxO metabolic control.
Cell metabolism (2008-02-06)
Michel L Tremblay, Vincent Giguère
ABSTRACT
FoxO transcription factors contribute to cardiac muscle remodeling and insulin signaling, but how they link insulin resistance and maladaptive heart hypertrophy remains unknown. A new study by Ni et al. (2007) shows that sustained activation of FoxO1 or FoxO3 in cardiomyocytes selectively enhances the activity of Akt/PKB and reduces insulin signaling through inhibition of calcineurin and PP2A.
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, Bacterial Glycerol Stock, Phosphatases
Sigma-Aldrich
MISSION® shRNA Mouse Gene Family Set, Bacterial Glycerol Stock, Phosphatases
Sigma-Aldrich
MISSION® shRNA Mouse Gene Family Set, Lentiviral Particles, Phosphatases
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, DNA, Phosphatases
Sigma-Aldrich
MISSION® shRNA Mouse Gene Family Set, DNA, Phosphatases
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, Lentiviral Particles, Phosphatases