Merck
  • A simple and efficient method to transfect small interference RNA into bovine SCNT embryos.
A simple and efficient method to transfect small interference RNA into bovine SCNT embryos.
Theriogenology (2015-07-22)
Hui Zhang, LiJun Wang, WenZhe Li, QingFu Mao, YongSheng Wang, Qian Li, Song Hua, Yong Zhang
ABSTRACT
RNA interference is an important tool to study the gene function. Microinjection and electroporation are usually used to transfer DNA, small interference RNA (siRNA), morpholinos, and protein into oocytes or embryos. This study used a simple and effective method to transfect siRNA into bovine somatic cell nuclear transfer (SCNT) embryos. In this method, siRNA transfection and electrofusion of SCNT were combined. A pair of platinum microelectrodes was used during SCNT to complete electrofusion. A CY3-labeled siRNA-targeted DNA methyltransferase-1 (DNMT1) was chosen to verify the siRNA transfection efficiency of this approach. First, a suitable concentration of siRNA was mixed with Zimmermann's fusion medium. Reconstructed embryos were then added into the microdrops of the mixed fusion medium to simultaneously transfect the siRNA and electrofuse the SCNT embryos. Our results showed that transfecting DNMT1 siRNA via the proposed method caused obvious CY3 fluorescence and significant downregulation of DNMT1 messenger RNA, DNMT1 protein, and global DNA methylation levels in the SCNT embryos. Meanwhile, the survival rate after electrofusion (90.4% vs. 89.4% vs. 89.1%, P > 0.05) and developmental rates of the SCNT embryos (72.8% vs. 74.9% vs. 72.4%, P > 0.05; 29.7% vs. 31.7% vs. 29.7%, P > 0.05) were not significantly affected. In summary, siRNAs were effectively transfected into the SCNT embryos via the proposed method and exert their functions, and the normal development of preimplantation SCNT embryos was not affected by the method used.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Dnmt1
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting human DNMT1
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Fluorescein, for fluorescence, free acid