Merck
  • Preparation of asparagine-linked monoglucosylated high-mannose-type oligosaccharide from egg yolk.
Preparation of asparagine-linked monoglucosylated high-mannose-type oligosaccharide from egg yolk.
Carbohydrate research (2015-05-15)
Ning Wang, Akira Seko, Yoichi Takeda, Yukishige Ito
ABSTRACT
Monoglucosylated high-mannose-type glycan (Glc1Man9GlcNAc2: G1M9) is well-known as a key glycoform in the glycoprotein folding process, which is specifically recognized by lectin chaperones calnexin (CNX) and calreticulin (CRT) in the endoplasmic reticulum (ER). In this work, we developed an efficient method for the preparation of G1M9-Asn. The G1M9-Asn was obtained from the IgY-rich fraction derived from hen egg yolk by the digestion with pronase. The α-amino group of asparagine in G1M9-Asn was protected with the 9-fluorenylmethyloxycarbonyl (Fmoc) group and the labeled glycans were subsequently purified using high performance liquid chromatography (HPLC). This method will provide useful substrates for analysis of the glycoprotein folding cycle in the ER.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Fmoc N-hydroxysuccinimide ester, ≥98.0% (HPLC)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Methanol, NMR reference standard
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
2,5-Dihydroxybenzoic acid, matrix substance for MALDI-MS, >99.0% (HPLC)
Sigma-Aldrich
2,5-Dihydroxybenzoic acid, 98%
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.