Merck
  • Lysophosphatidylserine has Bilateral Effects on Macrophages in the Pathogenesis of Atherosclerosis.
Lysophosphatidylserine has Bilateral Effects on Macrophages in the Pathogenesis of Atherosclerosis.
Journal of atherosclerosis and thrombosis (2014-12-03)
Masako Nishikawa, Makoto Kurano, Hitoshi Ikeda, Junken Aoki, Yutaka Yatomi
ABSTRACT
Lysophospholipids, particularly sphingosine 1-phosphate and lysophosphatidic acid, are known to be involved in the pathogenesis of atherosclerosis; however, the role of lysophosphatidylserine (LysoPS) in the onset of atherosclerotic diseases remains uncertain. We investigated the effects of LysoPS on the uptake of oxidized low-density lipoprotein (oxLDL) and the modulation of inflammatory mediators and ER stress utilizing RAW264.7 cells and mouse peritoneal macrophages (MPMs). We found that LysoPS augmented cholesterol accumulation in both models. Consistent with these findings, LysoPS increased the expression of scavenger receptors (CD36, MSR1, LOX1 and TLR4). Regarding the involvement of these lipids in inflammation, LysoPS significantly decreased the expression of inflammatory mediators in lipopolysaccharide (LPS)-treated RAW264.7 cells and MPMs. LysoPS also attenuated ER stress in LPS-untreated RAW264.7 cells. The expression patterns of LysoPS receptors differed considerably among the LPS-untreated RAW264.7 cells, LPS-treated RAW264.7 cells and MPMs. LysoPS may have proatherosclerotic properties in the setting of foam cell formation as well as antiatherosclerotic effects on inflammation in macrophages.
Product Description
Avanti
18:1 Lyso PS, Avanti Research - A Croda Brand
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, NMR reference standard
Avanti
18:1 Lyso PS, 1-oleoyl-2-hydroxy-sn-glycero-3-phospho-L-serine (sodium salt), chloroform
Avanti
18:0 Lyso PS, 1-stearoyl-2-hydroxy-sn-glycero-3-phospho-L-serine (sodium salt), powder
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.