Merck
  • A HAMP promoter bioassay system for identifying chemical compounds that modulate hepcidin expression.
A HAMP promoter bioassay system for identifying chemical compounds that modulate hepcidin expression.
Experimental hematology (2015-01-31)
Hiroshi Kawabata, Tatsuki Uchiyama, Soichiro Sakamoto, Junya Kanda, Shinya Oishi, Nobutaka Fujii, Naohisa Tomosugi, Norimitsu Kadowaki, Akifumi Takaori-Kondo
ABSTRACT
Hepcidin is the central regulator of systemic iron homeostasis; dysregulation of hepcidin expression causes various iron metabolic disorders, including hereditary hemochromatosis and anemia of inflammation. To identify molecules that modulate hepcidin expression, we developed a bioassay system for hepcidin gene (HAMP) promoter activity by stable transfection of Hep3B hepatoma cells with an expression plasmid in which EGFP was linked to a 2.5-kb human HAMP promoter. Interleukin 6, bone morphogenetic protein 6 (BMP-6), and oncostatin M, well-characterized stimulators of the HAMP promoter, strongly enhanced the green fluorescence intensity of these cells. Dorsomorphin, heparin, and cobalt chloride, known inhibitors of hepcidin expression, significantly suppressed green fluorescence intensity, and these inhibitory effects were more prominent when the cells were stimulated with BMP-6. Employing this system, we screened 1,280 biologically active small molecules and found several candidate inhibitors of hepcidin expression. Apomorphine, benzamil, etoposide, CGS-15943, kenpaullone, and rutaecarpine (all at 10 μmol/L) significantly inhibited hepcidin mRNA expression by Hep3B cells without affecting cell viability. CGS-15943 was the strongest suppressor of BMP-6-induced hepcidin-25 secretion in these cells. We conclude that our newly developed hepcidin promoter bioassay system is useful for identifying and evaluating compounds that modulate hepcidin expression.
Sigma-Aldrich
Interleukin-6 human, IL-6, recombinant, expressed in HEK 293 cells, suitable for cell culture
Sigma-Aldrich
R-(−)-Apomorphine hydrochloride hemihydrate, calcined, ≥98% (with NaOH, titration)
Sigma-Aldrich
Interleukin-6 human, IL-6, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Iron(III) citrate, technical grade
Sigma-Aldrich
Ferric citrate, BioReagent, suitable for cell culture
Sigma-Aldrich
EPO human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)
SAFC
Ferric citrate
Sigma-Aldrich
Interleukin-6 human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
BMP-6 human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture
Sigma-Aldrich
Bone Morphogenetic Protein 6 human, >95% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture