Merck
Nanoscale Nitrogen Doping in Silicon by Self-Assembled Monolayers.
Scientific reports (2015-08-01)
Bin Guan, Hamidreza Siampour, Zhao Fan, Shun Wang, Xiang Yang Kong, Abdelmadjid Mesli, Jian Zhang, Yaping Dan
ABSTRACT
This Report presents a nitrogen-doping method by chemically forming self-assembled monolayers on silicon. Van der Pauw technique, secondary-ion mass spectroscopy and low temperature Hall effect measurements are employed to characterize the nitrogen dopants. The experimental data show that the diffusion coefficient of nitrogen dopants is 3.66 × 10(-15) cm(2) s(-1), 2 orders magnitude lower than that of phosphorus dopants in silicon. It is found that less than 1% of nitrogen dopants exhibit electrical activity. The analysis of Hall effect data at low temperatures indicates that the donor energy level for nitrogen dopants is located at 189 meV below the conduction band, consistent with the literature value.
Sigma-Aldrich
Undecylenic acid, natural, ≥97%, FG
Sigma-Aldrich
Argon, ≥99.998%
Sigma-Aldrich
10-Undecenoic acid, 98%
Sigma-Aldrich
Argon-40Ar, 99.95 atom %