Merck
  • Laser-scanning cytometry can quantify human adipocyte browning and proves effectiveness of irisin.
Laser-scanning cytometry can quantify human adipocyte browning and proves effectiveness of irisin.
Scientific reports (2015-07-28)
Endre Kristóf, Quang-Minh Doan-Xuan, Péter Bai, Zsolt Bacso, László Fésüs
ABSTRACT
Laser-scanning cytometry is presented as a tool allowing population scale analysis of ex vivo human brown adipogenic differentiation. It combines texture analysis and detection of Ucp1 protein content in single brown adipocytes of mixed cell populations with gene expression pattern and functional characteristics of browning. Using this method we could validate mouse data in human samples demonstrating the effectiveness of irisin to induce "beige" differentiation of subcutaneous white adipocytes.
Sigma-Aldrich
Bone Morphogenetic Protein 7 human, >95% (SDS-PAGE), recombinant, expressed in CHO cells, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture