Merck
  • Targeting Homologous Recombination in Notch-Driven C. elegans Stem Cell and Human Tumors.
Targeting Homologous Recombination in Notch-Driven C. elegans Stem Cell and Human Tumors.
PloS one (2015-06-30)
Xinzhu Deng, David Michaelson, Jason Tchieu, Jin Cheng, Diana Rothenstein, Regina Feldman, Sang-gyu Lee, John Fuller, Adriana Haimovitz-Friedman, Lorenz Studer, Simon Powell, Zvi Fuks, E Jane Albert Hubbard, Richard Kolesnick
ABSTRACT
Mammalian NOTCH1-4 receptors are all associated with human malignancy, although exact roles remain enigmatic. Here we employ glp-1(ar202), a temperature-sensitive gain-of-function C. elegans NOTCH mutant, to delineate NOTCH-driven tumor responses to radiotherapy. At ≤20°C, glp-1(ar202) is wild-type, whereas at 25°C it forms a germline stem cell⁄progenitor cell tumor reminiscent of human cancer. We identify a NOTCH tumor phenotype in which all tumor cells traffic rapidly to G2⁄M post-irradiation, attempt to repair DNA strand breaks exclusively via homology-driven repair, and when this fails die by mitotic death. Homology-driven repair inactivation is dramatically radiosensitizing. We show that these concepts translate directly to human cancer models.
Sigma-Aldrich
MISSION® esiRNA, targeting human XRCC2
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
MISSION® esiRNA, targeting human RAD51
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Tributylphosphine solution, 200 mM (in N-methyl-2-pyrrolidinone), liquid
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Xrcc2