Merck
Coordinated expression of DNAM-1 and LFA-1 in educated NK cells.
Journal of immunology (Baltimore, Md. : 1950) (2015-04-01)
Monika Enqvist, Eivind Heggernes Ask, Elin Forslund, Mattias Carlsten, Greger Abrahamsen, Vivien Béziat, Sandra Andersson, Marie Schaffer, Anne Spurkland, Yenan Bryceson, Björn Önfelt, Karl-Johan Malmberg
ABSTRACT
The functional capacity of NK cells is dynamically tuned by integrated signals from inhibitory and activating cell surface receptors in a process termed NK cell education. However, the understanding of the cellular and molecular mechanisms behind this functional tuning is limited. In this study, we show that the expression of the adhesion molecule and activation receptor DNAX accessory molecule 1 (DNAM-1) correlates with the quantity and quality of the inhibitory input by HLA class I-specific killer cell Ig-like receptors and CD94/NKG2A as well as with the magnitude of functional responses. Upon target cell recognition, the conformational state of LFA-1 changed in educated NK cells, associated with rapid colocalization of both active LFA-1 and DNAM-1 at the immune synapse. Thus, the coordinated expression of LFA-1 and DNAM-1 is a central component of NK cell education and provides a potential mechanism for controlling cytotoxicity by functionally mature NK cells.
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥95.0% (HPLC)
Sigma-Aldrich
Abberior® STAR 635, phalloidin, for STED application
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)