Merck
  • Strontium ranelate effect on bone mineral density is modified by previous bisphosphonate treatment.
Strontium ranelate effect on bone mineral density is modified by previous bisphosphonate treatment.
SpringerPlus (2014-12-19)
Lucas R Brun, Ana M Galich, Eduardo Vega, Helena Salerni, Laura Maffei, Valeria Premrou, Pablo R Costanzo, Marcelo A Sarli, Paula Rey, María S Larroudé, María S Moggia, María L Brance, Ariel Sánchez
ABSTRACT
The aim of this study was to evaluate the effect of strontium ranelate (SrR) on bone mineral density (BMD) and bone turnover markers after 1 year of treatment. Additionally, the effect of SrR in bisphosphonate-naïve patients (BP-naïve) compared to patients previously treated with bisphosphonates (BP-prior) was analyzed. This retrospective study included 482 postmenopausal women treated with SrR (2 g/day) for 1 year in ten Argentine centers; 41 patients were excluded due to insufficient data, while 441 were included. Participants were divided according to previous bisphosphonate treatment in two groups: BP-naïve (n = 87) and BP-prior (n = 350). Data are expressed as mean ± SEM. After 1 year of treatment with SrR the bone formation markers total alkaline phosphatase and osteocalcin were increased (p < 0.0001), while the bone resorption marker s-CTX was decreased (p = 0.0579). Also increases in BMD at the lumbar spine (LS, 3.73%), femoral neck (FN, 2.00%) and total hip (TH, 1.54%) [p < 0.0001] were observed. These increments were significant (p < 0.0001) both among BP-naïve and BP-prior patients. Interestingly, the change in BMD after 1 year of SrR treatment was higher in BP-naïve patients: LS: BP-naïve = 4.58 ± 0.62%; BP-prior = 3.45 ± 0.28% (p = 0.078). FN: BP-naïve = 2.79 ± 0.56%; BP-prior = 2.13 ± 0.29% (p = 0.161). TH: BP-naïve = 3.01 ± 0.55%; BP-prior = 1.22 ± 0.27% (p = 0.0006). SrR treatment increased BMD and bone formation markers and decreased a bone resorption marker in the whole group, with better response in BP-naïve patients.
Sigma-Aldrich
β-Glycerophosphate disodium salt hydrate, BioUltra, suitable for cell culture, suitable for plant cell culture, ≥99% (titration)
Sigma-Aldrich
Glycerol phosphate disodium salt hydrate, isomeric mixture
Sigma-Aldrich
β-Glycerophosphate disodium salt hydrate, ≤1.0 mol % L-α-isomer