Merck
  • Development of Sensitive and Specific Analysis of Vildagliptin in Pharmaceutical Formulation by Gas Chromatography-Mass Spectrometry.
Development of Sensitive and Specific Analysis of Vildagliptin in Pharmaceutical Formulation by Gas Chromatography-Mass Spectrometry.
Journal of analytical methods in chemistry (2015-12-19)
Ebru Uçaktürk
ABSTRACT
A sensitive and selective gas chromatography-mass spectrometry (GC-MS) method was developed and fully validated for the determination of vildagliptin (VIL) in pharmaceutical formulation. Prior to GC-MS analysis, VIL was efficiently derivatized with MSTFA/NH4I/β-mercaptoethanol at 60°C for 30 min. The obtained O-TMS derivative of VIL was detected by selected ion monitoring mode using the diagnostic ions m/z 223 and 252. Nandrolone was chosen as internal standard. The GC-MS method was fully validated by the following validation parameters: limit of detection (LOD) and quantitation (LOQ), linearity, precision, accuracy, specificity, stability, robustness, and ruggedness. LOD and LOQ were found to be 1.5 and 3.5 ng mL(-1), respectively. The GC-MS method is linear in the range of 3.5-300 ng mL(-1). The intra- and interday precision values were less than ≤3.62%. The intra- and interday accuracy values were found in the range of -0.26-2.06%. Finally, the GC-MS method was successfully applied to determine VIL in pharmaceutical formulation.
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Ammonium iodide, 99.999% trace metals basis
Supelco
N-Methyl-bis(trifluoroacetamide), for GC derivatization, LiChropur, ≥97.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Ammonium iodide, ACS reagent, ≥99%
Sigma-Aldrich
Acetic anhydride, 99.5%
Sigma-Aldrich
Acetic anhydride, Arxada quality, ≥99.5% (GC)
Sigma-Aldrich
Acetic anhydride, ACS reagent, ≥98.0%
Sigma-Aldrich
Acetic anhydride, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic anhydride, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99% (GC)