Merck
  • Lipopolysaccharides, cytokines, and nitric oxide affect secretion of prostaglandins and leukotrienes by bovine mammary gland during experimentally induced mastitis in vivo and in vitro.
Lipopolysaccharides, cytokines, and nitric oxide affect secretion of prostaglandins and leukotrienes by bovine mammary gland during experimentally induced mastitis in vivo and in vitro.
Domestic animal endocrinology (2015-05-04)
K K Piotrowska-Tomala, M M Bah, K Jankowska, K Lukasik, P Warmowski, A M Galvao, D J Skarzynski
ABSTRACT
The aim of the study was to determine the effects of lipopolysaccharide (LPS), tumor necrosis factor alpha (TNF), interleukin-1-alpha (IL-1α), and nitric oxide donor (NONOate) on both in vivo and in vitro secretion of prostaglandin (PG)E2, PGF2α, leukotriene (LT)B4, and LTC4 by the bovine mammary gland. In the first experiment, tissues isolated from the teat cavity and lactiferous sinus were treated in vitro with LPS (10 ng/mL), TNF (10 ng/mL), IL-1α (10 ng/mL), NONOate (10(-4) M), and the combination of TNF + IL-1α + NONOate for 4 or 8 h. PGE2 or PGF2α secretion was stimulated by all treatments (P < 0.05) excepting NONOate alone, which did not stimulate PGF2α secretion. Moreover, all factors increased LTB4 and LTC4 secretion (P < 0.05). In the second experiment, mastitis was experimentally mimicked in vivo by repeated (12 h apart) intramammary infusions (5 mL) of (1) sterile saline; (2) 250-μg LPS; (3) 1-μg/mL TNF; (4) 1-μg/mL IL-1α; (5) 12.8-μg/mL NONOate; and (6) TNF + IL-1α + NONOate into 2 udder quarters. All infused factors changed PGE2, 13,14-dihydro,15-keto-PGF2α, and LT concentrations in blood plasma collected from the caudal vena cava, the caudal superficial epigastric (milk) vein, the jugular vein, and the abdominal aorta (P < 0.05). In summary, LPS and other inflammatory mastitis mediators modulate PG and LT secretion by bovine mammary gland in both in vivo and in vitro studies.
Sigma-Aldrich
TNF-α human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, microbiologically tested, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
IL-10 human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine and 15 mM HEPES, without sodium bicarbonate and phenol red, powder, suitable for cell culture
Sigma-Aldrich
Tumor Necrosis Factor-α human, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
Interleukin-10 human, ≥97% (SDS-PAGE), recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Gentamicin solution, 50 mg/mL in deionized water, liquid, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Antibiotic Antimycotic Solution (100×), Stabilized, with 10,000 units penicillin, 10 mg streptomycin and 25 μg amphotericin B per mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
1,3-Propanediol, 98%
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
IL-10 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)