Merck
  • Determination of four forms of vitamin B12 and other B vitamins in seawater by liquid chromatography/tandem mass spectrometry.
Determination of four forms of vitamin B12 and other B vitamins in seawater by liquid chromatography/tandem mass spectrometry.
Rapid communications in mass spectrometry : RCM (2014-10-11)
Katherine R Heal, Laura Truxal Carlson, Allan H Devol, E Virginia Armbrust, James W Moffett, David A Stahl, Anitra E Ingalls
ABSTRACT
Vitamin B(12) is an essential nutrient for more than half of surveyed marine algae species, but methods for directly measuring this important cofactor in seawater are limited. Current mass spectrometry methods do not quantify all forms of B(12), potentially missing a significant portion of the B(12) pool. We present a method to measure vitamins B(1), B(2), B(6), B(7) and four forms of B(12) dissolved in seawater. The method entails solid-phase extraction, separation by ultra-performance liquid chromatography, and detection by triple-quadrupole tandem mass spectrometry using stable-isotope-labeled internal standards. We demonstrated the use of this method in the environment by analyzing B(12) concentrations at different depths in the Hood Canal, part of the Puget Sound estuarine system in Washington State. Recovery of vitamin B(12) forms during the preconcentration steps was >71% and the limits of detection were <0.275 pM in seawater. Standard addition calibration curves in three different seawater matrices were used to determine analytical response and to quantify samples from the environment. Hydroxocobalamin was the main form of B(12) in seawater at our field site. We developed a method for quantifying four forms of B(12) in seawater by liquid chromatography/mass spectrometry with the option of simultaneous analysis of vitamins B(1), B(2), B(6), and B(7). We validated the method and demonstrated its application in the field.
Product Description
Supelco
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Thiamine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Riboflavin, European Pharmacopoeia (EP) Reference Standard
USP
Thiamine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Supelco
Thiamine Hydrochloride (B1), analytical standard
Sigma-Aldrich
Thiamine hydrochloride, ≥98%, FCC, FG
Sigma-Aldrich
Thiamine hydrochloride, tested according to Ph. Eur.
Sigma-Aldrich
Thiamine hydrochloride, reagent grade, ≥99% (HPLC)
Sigma-Aldrich
Thiamine hydrochloride, meets USP testing specifications
Sigma-Aldrich
Thiamine hydrochloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Thiamine hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Riboflavin, United States Pharmacopeia (USP) Reference Standard
Cyanocobalamin, British Pharmacopoeia (BP) Assay Standard
Riboflavin for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Cyanocobalamin, pharmaceutical secondary standard, certified reference material
Supelco
Riboflavin (B2), analytical standard
Supelco
Cyanocobalamin (B12), analytical standard
Sigma-Aldrich
Cyanocobalamin, tested according to Ph. Eur.
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Sigma-Aldrich
Pyridoxine, ≥98%
Cyanocobalamin, European Pharmacopoeia (EP) Reference Standard