Merck
  • Co-expression of PKM2 and TRIM35 predicts survival and recurrence in hepatocellular carcinoma.
Co-expression of PKM2 and TRIM35 predicts survival and recurrence in hepatocellular carcinoma.
Oncotarget (2015-01-13)
Zhiao Chen, Xinyuan Lu, Zhichao Wang, Guangzhi Jin, Qifeng Wang, Di Chen, Taoyang Chen, Jinjun Li, Jia Fan, Wenming Cong, Qiang Gao, Xianghuo He
ABSTRACT
The identification of prognostic markers for hepatocellular carcinoma (HCC) is needed for clinical practice. Tripartite motif-containing 35 (TRIM35) is a tumor suppressor of HCC. TRIM35 inhibits phosphorylation of pyruvate kinase isoform M2 (PKM2), which is involved in aerobic glycolysis of cancer cells. We found that expression of PKM2 was significantly increased in HCC tissues. This overexpression of PKM2 was correlated with a high TNM stage and level of vascular invasion. Patients with HCC who were positive for PKM2 expression and negative for TRIM35 expression had shorter overall survival and time to recurrence than patients who were negative for PKM2 and positive for TRIM35. Furthermore, PKM2/TRIM35 combination was an independent and significant risk factor for recurrence and survival. In conclusion, PKM2 (+) and TRIM35 (-) contribute to the aggressiveness and poor prognosis of HCC. PKM2/TRIM35 expression could be a biomarker for the prognosis of HCC and target for cancer therapy.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Anti-TRIM35 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard