Merck
  • RETRACTED: How does high-concentration supplemental perioperative oxygen influence surgical outcomes after thyroid surgery? A prospective, randomized, double-blind, controlled, monocentric trial.
RETRACTED: How does high-concentration supplemental perioperative oxygen influence surgical outcomes after thyroid surgery? A prospective, randomized, double-blind, controlled, monocentric trial.
Journal of the American College of Surgeons (2015-04-05)
Mario Schietroma, Federica Piccione, Emanuela Marina Cecilia, Francesco Carlei, Giuseppe De Santis, Federico Sista, Gianfranco Amicucci
ABSTRACT
This article has been retracted: please see Elsevier Policy on Article Withdrawal (https://www.elsevier.com/about/our-business/policies/article-withdrawal). This article has been retracted at the request of the Editor-in-Chief. An independent statistical analysis based on the summary data tables and statistical results reported in the article confirmed that the statistical results are incorrect and the data do not support the conclusions of the article.
Sigma-Aldrich
Promethazine hydrochloride
Promethazine for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Promethazine hydrochloride, vial of 25 mg, certified reference material, Cerilliant®
Promethazine hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Promethazine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Supelco
Promethazine Hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Atracurium for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Atropine sulfate salt monohydrate, ≥97% (TLC), crystalline
USP
Atropine sulfate, United States Pharmacopeia (USP) Reference Standard
USP
Atracurium besylate, United States Pharmacopeia (USP) Reference Standard
Atropine sulfate, European Pharmacopoeia (EP) Reference Standard
Supelco
Atropine Sulfate, Pharmaceutical Secondary Standard; Certified Reference Material
Atracurium for impurity F identification, European Pharmacopoeia (EP) Reference Standard
Atracurium besylate, European Pharmacopoeia (EP) Reference Standard