Merck
  • MicroRNA-155 is required for clearance of Streptococcus pneumoniae from the nasopharynx.
MicroRNA-155 is required for clearance of Streptococcus pneumoniae from the nasopharynx.
Infection and immunity (2014-08-27)
Chris P Verschoor, Michael G Dorrington, Kyle E Novakowski, Julie Kaiser, Katherine Radford, Parameswaran Nair, Varun Anipindi, Charu Kaushic, Michael G Surette, Dawn M E Bowdish
ABSTRACT
Pneumonia caused by Streptococcus pneumoniae is a major cause of death and an economic burden worldwide. S. pneumoniae is an intermittent colonizer of the human upper respiratory tract, and the ability to control asymptomatic colonization determines the likelihood of developing invasive disease. Recognition of S. pneumoniae by resident macrophages via Toll-like receptor 2 (TLR-2) and the macrophage receptor with collagenous structure (MARCO) and the presence of interleukin-17 (IL-17)-secreting CD4(+) T cells are required for macrophage recruitment and bacterial clearance. Despite the fact that the primary cellular effectors needed for bacterial clearance have been identified, much of the underlying regulatory mechanisms are unknown. Herein, we demonstrate that the small, noncoding RNA microRNA-155 (mir-155) is critical for the effective clearance of S. pneumoniae. Our studies show that mir-155-deficient mice maintain the ability to prevent acute invasive pneumococcal infection but have significantly higher bacterial burdens following colonization, independently of macrophage recognition by TLR-2, MARCO expression, or bactericidal capacity. The observed defects in bacterial clearance parallel reduced IL-17A and gamma interferon CD4(+) T-cell responses in vivo, lower IL-17A mRNA levels in the nasopharynx, and a reduced capacity to induce Th17 cell polarization. Given that knockout mice are also limited in the capacity to generate high-titer S. pneumoniae-specific antibodies, we conclude that mir-155 is a critical mediator of the cellular effectors needed to clear primary and secondary S. pneumoniae colonizations.
Sigma-Aldrich
Rat IFN-γ ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Cotton Rat Ifng / Interferon Gamma ELISA Kit
Sigma-Aldrich
Bovine IFNG / Interferon Gamma ELISA Kit, for serum, plasma and cell culture supernatants
Sigma-Aldrich
Mouse IFN-γ ELISA Kit, for serum, plasma, cell culture supernatant
Sigma-Aldrich
Mouse IFN-γ ELISA Kit, for cell and tissue lysates