Merck
  • Clonal diversity of Acinetobacter baumannii from diabetic patients in Saudi Arabian hospitals.
Clonal diversity of Acinetobacter baumannii from diabetic patients in Saudi Arabian hospitals.
Journal of medical microbiology (2014-08-12)
Abdulrahman A Alsultan, Elsayed Aboulmagd, Benjamin A Evans, Sebastian G B Amyes
ABSTRACT
Carbapenem-resistant Acinetobacter baumannii (CR-AB) represents a major health-care problem, causing high rates of morbidity and mortality. This study investigated the clonality of CR-AB isolated from diabetic patients from different regions in Saudi Arabia, as well as the relatedness of the β-lactamase genes. A total of 64 non-repetitive CR-AB clinical isolates were collected from 16 different regions in Saudi Arabia from intensive care patients. Isolates were identified phenotypically by the Vitek 2 compact system and genotypically by amplification of the blaOXA-51-like gene. The target sequences were amplified by PCR and the clonal diversity of the isolates was explored by PFGE. Resistance studies revealed that the prevalence of imipenem and meropenem resistance was 92% and 96%, respectively, while the vast majority of the isolates were susceptible to tigecycline and colistin. In addition, blaVIM and blaOXA-23 were the most prevalent genes in the isolates under investigation, while ISAba1 was the most dominant insertion sequence. PFGE results showed 13 clusters; clone H was dominant, comprising 20 isolates from four hospitals, followed by clones C and F, comprising 11 isolates each from three and six hospitals, respectively. Moreover, the current study signified the clonal diversity of CR-AB in Saudi Arabia and showed the ability of some clones to infect patients in many different cities.
Product Description
Supelco
Isopropyl myristate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Sigma-Aldrich
Isopropyl myristate, ≥98%
Sigma-Aldrich
Isopropyl myristate, ≥90% (GC)
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
Isopropyl myristate, 98%
USP
Meropenem, United States Pharmacopeia (USP) Reference Standard
USP
Isopropyl myristate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethidium bromide, BioReagent, for molecular biology, powder
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)