Merck
  • Inhibition of miR-15a Promotes BDNF Expression and Rescues Dendritic Maturation Deficits in MeCP2-Deficient Neurons.
Inhibition of miR-15a Promotes BDNF Expression and Rescues Dendritic Maturation Deficits in MeCP2-Deficient Neurons.
Stem cells (Dayton, Ohio) (2015-02-03)
Yu Gao, Juan Su, Weixiang Guo, Eric D Polich, Daniel P Magyar, Yina Xing, Hongda Li, Richard D Smrt, Qiang Chang, Xinyu Zhao
ABSTRACT
In both the embryonic and adult brain, a critical step in neurogenesis is neuronal maturation. Deficiency of MeCP2 leads to Rett syndrome, a severe neurodevelopmental disorder. We have previously shown that MeCP2 plays critical roles in the maturation step of new neurons during neurogenesis. MeCP2 is known to regulate the expression of brain-derived neurotrophic factor (BDNF), a potent neurotrophic factor for neuronal maturation. Nevertheless, how MeCP2 regulates BDNF expression and how MeCP2 deficiency leads to reduced BDNF expression remain unclear. Here, we show that MeCP2 regulates the expression of a microRNA, miR-15a. We find that miR-15a plays a significant role in the regulation of neuronal maturation. Overexpression of miR-15a inhibits dendritic morphogenesis in immature neurons. Conversely, a reduction in miR-15a has the opposite effect. We further show that miR-15a regulates expression levels of BDNF, and exogenous BDNF could partially rescue the neuronal maturation deficits resulting from miR-15a overexpression. Finally, inhibition of miR-15a could rescue neuronal maturation deficits in MeCP2-deficient adult-born new neurons. These results demonstrate a novel role for miR-15a in neuronal development and provide a missing link in the regulation of BDNF by MeCP2.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Sigma-Aldrich
MISSION® esiRNA, targeting human PRR14
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Sodium chloride solution, 5 M