Merck
  • High throughput quantification of prohibited substances in plasma using thin film solid phase microextraction.
High throughput quantification of prohibited substances in plasma using thin film solid phase microextraction.
Journal of chromatography. A (2014-12-03)
Nathaly Reyes-Garcés, Barbara Bojko, Janusz Pawliszyn
ABSTRACT
Simple, fast and efficient sample preparation approaches that allow high-throughput isolation of various compounds from complex matrices are highly desired in bioanalysis. Particularly sought are methods that can, without sacrificing time, easily remove matrix interferences capable of inducing ionization suppression/enhancement, or causing detrimental effects in instrumental performance. In this work, an automated high-throughput sample preparation method using thin film solid phase microextraction (SPME) for the analysis of multiple prohibited substances in plasma is proposed. A biocompatible SPME extraction phase made of hydrophilic-lipophilic balance particles immobilized with polyacrylonitrile (PAN) demonstrated satisfactory extraction capabilities for 25 compounds of a wide range of polarities (logP from -2 to 6.8). Due to the well-known biocompatible characteristics of PAN-based SPME coatings, minimum sample handling was required. Experimental conditions for pre-conditioning, extraction, wash and desorption were carefully optimized for the proposed method. By taking full advantage of the 96 thin film handling capability of the automated system, a preparation time of approximately 1.5min per sample can be achieved. Satisfactory results in terms of absolute matrix effects were found for the majority of the studied analytes, given that 24 out of 25 compounds exhibited values in the range of 100 and 120%. The method was validated in terms of linearity (R(2)>0.99), inter and intra-day accuracy (85-130%) and precision (<20%) and limits of quantitation (0.25-10ngmL(-1) for most compounds).
Product Description
Supelco
Furosemide, Pharmaceutical Secondary Standard; Certified Reference Material
Furosemide, European Pharmacopoeia (EP) Reference Standard
USP
Furosemide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Exemestane, ≥98% (HPLC)
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, for use with autosampler, needle size 23 ga, metal alloy fiber
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Supelco
SPME fiber assortment kit 4, for use with autosampler, size 24 ga
Supelco
SPME fiber assortment kit 2, for use with autosampler, needle size 23 ga
Supelco
SPME StableFlex fiber assortment kit, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assortment kit 2, for use with autosampler, size 24 ga
Supelco
SPME StableFlex fiber assortment kit, for use with manual holder
Supelco
SPME fiber assortment kit 1, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assortment kit 1, for use with autosampler, needle size 24 ga
Supelco
SPME fiber assortment kit 4, for use with manual holder, size 24 ga
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assortment kit 2, for use with manual holder, needle size 24 ga
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Salbutamol
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Strychnine
Supelco
SPME fiber assortment kit 5, needle size 23 ga, for use with autosampler
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%