Merck
  • Adhesion of Staphylococcus aureus to the vessel wall under flow is mediated by von Willebrand factor-binding protein.
Adhesion of Staphylococcus aureus to the vessel wall under flow is mediated by von Willebrand factor-binding protein.
Blood (2014-06-22)
Jorien Claes, Thomas Vanassche, Marijke Peetermans, Laurens Liesenborghs, Christophe Vandenbriele, Karen Vanhoorelbeke, Dominique Missiakas, Olaf Schneewind, Marc F Hoylaerts, Ruth Heying, Peter Verhamme
ABSTRACT
Adhesion of Staphylococcus aureus to blood vessels under shear stress requires von Willebrand factor (VWF). Several bacterial factors have been proposed to interact with VWF, including VWF-binding protein (vWbp), a secreted coagulase that activates the host's prothrombin to generate fibrin. We measured the adhesion of S aureus Newman and a vWbp-deficient mutant (vwb) to VWF, collagen, and activated endothelial cells in a microparallel flow chamber. In vivo adhesion of S aureus was evaluated in the mesenteric circulation of wild-type (WT) and VWF-deficient mice. We found a shear-dependent increase in adhesion of S aureus to the (sub)endothelium that was dependent on interactions between vWbp and the A1-domain of VWF. Adhesion was further enhanced by coagulase-mediated fibrin formation that clustered bacteria and recruited platelets into bacterial microthrombi. In vivo, deficiency of vWbp or VWF as well as inhibition of coagulase activity reduced S aureus adhesion. We conclude that vWbp contributes to vascular adhesion of S aureus through 2 independent mechanisms: shear-mediated binding to VWF and activation of prothrombin to form S aureus-fibrin-platelet aggregates.
Sigma-Aldrich
5(6)-Carboxyfluorescein, Dye content 90 %
Pricing and availability is not currently available.
Sigma-Aldrich
5(6)-Carboxyfluorescein, suitable for fluorescence, BioReagent, ≥95% (HPLC)
Pricing and availability is not currently available.