Merck
  • Genetic ablation of macrohistone H2A1 leads to increased leanness, glucose tolerance and energy expenditure in mice fed a high-fat diet.
Genetic ablation of macrohistone H2A1 leads to increased leanness, glucose tolerance and energy expenditure in mice fed a high-fat diet.
International journal of obesity (2005) (2014-05-23)
F Sheedfar, M Vermeer, V Pazienza, J Villarroya, F Rappa, F Cappello, G Mazzoccoli, F Villarroya, H van der Molen, M H Hofker, D P Koonen, M Vinciguerra
ABSTRACT
In the context of obesity, epigenetic mechanisms regulate cell-specific chromatin plasticity, perpetuating gene expression responses to nutrient excess. MacroH2A1, a variant of histone H2A, emerged as a key chromatin regulator sensing small nutrients during cell proliferation and differentiation. Mice genetically ablated for macroH2A1 (knockout (KO)) do not show overt phenotypes under a standard diet. Our objective was to analyse the in vivo role of macroH2A1 in response to nutritional excess. Twelve-week-old whole-body macroH2A1 KO male mice were given a high-fat diet (60% energy from lard) for 12 weeks until being killed, and examined for glucose and insulin tolerance, and for body fat composition. Energy expenditure was assessed using metabolic cages and by measuring the expression levels of genes involved in thermogenesis in the brown adipose tissue (BAT) or in adipogenesis in the visceral adipose tissue (VAT). Under a chow diet, macroH2A1 KO mice did not differ from their wild-type (WT) littermates for body weight, and for sensitivity to glucose or insulin. However, KO mice displayed decreased heat production (P<0.05), and enhanced total activity during the night (P<0.01). These activities related to protection against diet-induced obesity in KO mice, which displayed decreased body weight owing to a specific decrease in fat mass (P<0.05), increased tolerance to glucose (P<0.05), and enhanced total activity during the day (P<0.05), compared with WT mice. KO mice displayed increased expression of thermogenic genes (Ucp1, P<0.05; Glut4, P<0.05; Cox4, P<0.01) in BAT and a decreased expression of adipogenic genes (Pparγ, P<0.05; Fabp4, P<0.05; Glut4, P<0.05) in VAT compared with WT mice, indicative of augmented energy expenditure. Genetic eviction of macroH2A1 confers protection against diet-induced obesity and metabolic derangements in mice. Inhibition of macroH2A1 might be a helpful strategy for epigenetic therapy of obesity.
Product Description
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material