Merck
High rate and stable cycling of lithium metal anode.
Nature communications (2015-02-24)
Jiangfeng Qian, Wesley A Henderson, Wu Xu, Priyanka Bhattacharya, Mark Engelhard, Oleg Borodin, Ji-Guang Zhang
ABSTRACT
Lithium metal is an ideal battery anode. However, dendrite growth and limited Coulombic efficiency during cycling have prevented its practical application in rechargeable batteries. Herein, we report that the use of highly concentrated electrolytes composed of ether solvents and the lithium bis(fluorosulfonyl)imide salt enables the high-rate cycling of a lithium metal anode at high Coulombic efficiency (up to 99.1%) without dendrite growth. With 4 M lithium bis(fluorosulfonyl)imide in 1,2-dimethoxyethane as the electrolyte, a lithium|lithium cell can be cycled at 10 mA cm(-2) for more than 6,000 cycles, and a copper|lithium cell can be cycled at 4 mA cm(-2) for more than 1,000 cycles with an average Coulombic efficiency of 98.4%. These excellent performances can be attributed to the increased solvent coordination and increased availability of lithium ion concentration in the electrolyte. Further development of this electrolyte may enable practical applications for lithium metal anode in rechargeable batteries.
Sigma-Aldrich
Lithium hexafluorophosphate, battery grade, ≥99.99% trace metals basis
Supelco
1,2-Dimethoxyethane, analytical standard
Sigma-Aldrich
1,2-Dimethoxyethane, anhydrous, 99.5%, inhibitor-free
Sigma-Aldrich
Propylene carbonate, anhydrous, 99.7%
Sigma-Aldrich
Lithium hexafluorophosphate, 98%
Sigma-Aldrich
Propylene carbonate, ReagentPlus®, 99%
Sigma-Aldrich
1,2-Dimethoxyethane, ReagentPlus®, ≥99%, inhibitor-free
Sigma-Aldrich
Propylene carbonate, suitable for HPLC, 99.7%
Sigma-Aldrich
1,2-Dimethoxyethane, suitable for HPLC, 99.9%, inhibitor-free
Sigma-Aldrich
1,3-Dioxolane, anhydrous, contains ~75 ppm BHT as inhibitor, 99.8%
Sigma-Aldrich
Lithium-7Li hexafluorophosphate, 95 atom % 7Li, 98% (CP)
Sigma-Aldrich
1,3-Dioxolane, ReagentPlus®, contains ~75 ppm BHT as inhibitor, 99%