Merck
  • Comparative percutaneous permeation study using caffeine-loaded microemulsion showing low reliability of the frozen/thawed skin models.
Comparative percutaneous permeation study using caffeine-loaded microemulsion showing low reliability of the frozen/thawed skin models.
International journal of pharmaceutics (2014-05-29)
Amnon C Sintov, Igor Greenberg
ABSTRACT
The aim of this study was to explore the transdermal delivery potential of a new caffeine-containing microemulsion system. The skin permeability of caffeine (CAF) was measured in vitro using skin excised from three different animal species: rat, rabbit and pig. As shown, microemulsion containing 20% aqueous phase enhanced CAF permeation across fresh rat skin by one order of magnitude (Papp=8.2×10(-3) vs. 0.86×10(-3) cm/h; enhancement ratio=9.6). The permeability coefficient value, the cumulative permeation amount, and the percent of dose permeated after 24 h, decreased with the increase of water content from 60% to 80% in microemulsions due to the apparent increase in the droplet size. Importantly, differences were noted between caffeine transport rates across fresh and frozen/thawed pig skin whereas microemulsions delivered caffeine at similar rates across rat and rabbit skin, either fresh or frozen/thawed. It has been shown that the permeability of caffeine through frozen/thawed pig skin was abnormally high and was independent of its vehicle properties, i.e., its hydrophilic or lipophilic nature. It has been hypothesized that the reason for this abnormality is that porcine stratum corneum has a higher ceramide-to-cholesterol ratio compared to rat and rabbit skin. This unusual phenomenon observed in a non-freshly used porcine skin places a question mark on its suitability to in vitro evaluation of transdermal drug delivery systems.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Propylene carbonate, anhydrous, 99.7%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Melting point standard 235-237°C, analytical standard
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Propylene carbonate, suitable for HPLC, 99.7%
Isopropyl hexadecanoate, European Pharmacopoeia (EP) Reference Standard
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Propylene carbonate, ReagentPlus®, 99%
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Isopropyl palmitate, technical grade, 90%
Supelco
Isopropyl palmitate, Pharmaceutical Secondary Standard; Certified Reference Material