Merck
  • Thermally robust and porous noncovalent organic framework with high affinity for fluorocarbons and CFCs.
Thermally robust and porous noncovalent organic framework with high affinity for fluorocarbons and CFCs.
Nature communications (2014-10-14)
Teng-Hao Chen, Ilya Popov, Watchareeya Kaveevivitchai, Yu-Chun Chuang, Yu-Sheng Chen, Olafs Daugulis, Allan J Jacobson, Ognjen Š Miljanić
ABSTRACT
Metal-organic and covalent organic frameworks are porous materials characterized by outstanding thermal stability, high porosities and modular synthesis. Their repeating structures offer a great degree of control over pore sizes, dimensions and surface properties. Similarly precise engineering at the nanoscale is difficult to achieve with discrete molecules, since they rarely crystallize as porous structures. Here we report a small organic molecule that organizes into a noncovalent organic framework with large empty pores. This structure is held together by a combination of [N-H···N] hydrogen bonds between the terminal pyrazole rings and [π···π] stacking between the electron-rich pyrazoles and electron-poor tetrafluorobenzenes. Such a synergistic arrangement makes this structure stable to at least 250 °C and porous, with an accessible surface area of 1,159 m(2) g(-1). Crystals of this framework adsorb hydrocarbons, CFCs and fluorocarbons-the latter two being ozone-depleting substances and potent greenhouse species-with weight capacities of up to 75%.
Sigma-Aldrich
Lithium tert-butoxide solution, 1.0 M in THF
Sigma-Aldrich
Trityl chloride, purum, ≥97.0% (HPLC), ≥97.0% (AT)
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Supelco
Cyclohexane, analytical standard
Sigma-Aldrich
Trityl chloride, 97%
Sigma-Aldrich
Cyclohexane, anhydrous, 99.5%
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Cyclohexane, ACS reagent, ≥99%
Sigma-Aldrich
Cyclohexane, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Cyclohexane, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Cyclohexane, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
Cyclohexane, ACS reagent, ≥99%
Sigma-Aldrich
Cyclohexane, Laboratory Reagent, ≥99.8%
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Lithium tert-butoxide, 97%
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)