Merck
  • High dietary protein restores overreaching induced impairments in leukocyte trafficking and reduces the incidence of upper respiratory tract infection in elite cyclists.
High dietary protein restores overreaching induced impairments in leukocyte trafficking and reduces the incidence of upper respiratory tract infection in elite cyclists.
Brain, behavior, and immunity (2013-10-15)
Oliver C Witard, James E Turner, Sarah R Jackman, Arie K Kies, Asker E Jeukendrup, Jos A Bosch, Kevin D Tipton
ABSTRACT
The present study examined whether a high protein diet prevents the impaired leukocyte redistribution in response to acute exercise caused by a large volume of high-intensity exercise training. Eight cyclists (VO2max: 64.2±6.5mLkg(-1)min(-1)) undertook two separate weeks of high-intensity training while consuming either a high protein diet (3gkg(-1)proteinBM(-1)day(-1)) or an energy and carbohydrate-matched control diet (1.5gkg(-1)proteinBM(-1)day(-1)). High-intensity training weeks were preceded by a week of normal-intensity training under the control diet. Leukocyte and lymphocyte sub-population responses to acute exercise were determined at the end of each training week. Self-reported symptoms of upper-respiratory tract infections (URTI) were monitored daily by questionnaire. Undertaking high-intensity training with a high protein diet restored leukocyte kinetics to similar levels observed during normal-intensity training: CD8(+) TL mobilization (normal-intensity: 29,319±13,130cells/μL×∼165min vs. high-intensity with protein: 26,031±17,474cells/μL×∼165min, P>0.05), CD8(+) TL egress (normal-intensity: 624±264cells/μL vs. high-intensity with protein: 597±478cells/μL, P>0.05). This pattern was driven by effector-memory populations mobilizing (normal-intensity: 6,145±6,227cells/μL×∼165min vs. high-intensity with protein: 6,783±8,203cells/μL×∼165min, P>0.05) and extravastating from blood (normal-intensity: 147±129cells/μL vs. high-intensity with protein: 165±192cells/μL, P>0.05). High-intensity training while consuming a high protein diet was associated with fewer symptoms of URTI compared to performing high-intensity training with a normal diet (P<0.05). To conclude, a high protein diet might reduce the incidence of URTI in athletes potentially mediated by preventing training-induced impairments in immune-surveillance.
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Urea, European Pharmacopoeia (EP) Reference Standard
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Supelco
Urea, analytical standard