Merck
  • High warfarin sensitivity in carriers of CYP2C9*35 is determined by the impaired interaction with P450 oxidoreductase.
High warfarin sensitivity in carriers of CYP2C9*35 is determined by the impaired interaction with P450 oxidoreductase.
The pharmacogenomics journal (2013-12-11)
M-Y Lee, P Borgiani, I Johansson, F Oteri, S Mkrtchian, M Falconi, M Ingelman-Sundberg
ABSTRACT
Cytochrome P450 2C9 (CYP2C9) metabolizes many clinically important drugs including warfarin and diclofenac. We have recently reported a new allelic variant, CYP2C9*35, found in a warfarin hypersensitive patient with Arg125Leu and Arg144Cys mutations. Here, we have investigated the molecular basis for the functional consequences of these polymorphic changes. CYP2C9.1 and CYP2C9-Arg144Cys expressed in human embryonic kidney 293 cells effectively metabolized both S-warfarin and diclofenac in NADPH-dependent reactions, whereas CYP2C9-Arg125Leu or CYP2C9.35 were catalytically silent. However, when NADPH was replaced by a direct electron donor to CYPs, cumene hydroperoxide, hereby bypassing the CYP oxidoreductase (POR), all variant enzymes were active, indicating unproductive interactions between CYP2C9.35 and POR. In silico analysis revealed a decrease of the electrostatic potential of CYP2C9-Arg125Leu-POR interacting surface and the loss of stabilizing salt bridges between these proteins. In conclusion, our data strongly suggest that the Arg125Leu mutation in CYP2C9.35 prevents CYP2C9-POR interactions resulting in the absence of NADPH-dependent CYP2C9-catalyzed activity in vivo, thus influencing the warfarin sensitivity in the carriers of this allele.
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Supelco
4′-Hydroxydiclofenac, VETRANAL®, analytical standard
Sigma-Aldrich
Diclofenac sodium salt
Diclofenac for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Diclofenac sodium salt, analytical standard
Supelco
Diclofenac sodium salt, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phosphoric acid solution, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid solution, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
7-Hydroxywarfarin
Diclofenac sodium, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, puriss., ≥99%
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
USP
Diclofenac sodium, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Cumene hydroperoxide, technical grade, 80%
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Phosphoric acid, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, 85.0-88.0%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)