Merck
  • Sox9 is critical for suppression of neurogenesis but not initiation of gliogenesis in the cerebellum.
Sox9 is critical for suppression of neurogenesis but not initiation of gliogenesis in the cerebellum.
Molecular brain (2015-04-19)
Keng Ioi Vong, Crystal Kit Ying Leung, Richard R Behringer, Kin Ming Kwan
ABSTRACT
The high mobility group (HMG) family transcription factor Sox9 is critical for induction and maintenance of neural stem cell pool in the central nervous system (CNS). In the spinal cord and retina, Sox9 is also the master regulator that defines glial fate choice by mediating the neurogenic-to-gliogenic fate switch. On the other hand, the genetic repertoire governing the maintenance and fate decision of neural progenitor pool in the cerebellum has remained elusive. By employing the Cre/loxP strategy, we specifically inactivated Sox9 in the mouse cerebellum. Unexpectedly, the self-renewal capacity and multipotency of neural progenitors at the cerebellar ventricular zone (VZ) were not perturbed upon Sox9 ablation. Instead, the mutants exhibited an increased number of VZ-derived neurons including Purkinje cells and GABAergic interneurons. Simultaneously, we observed continuous neurogenesis from Sox9-null VZ at late gestation, when normally neurogenesis ceases to occur and gives way for gliogenesis. Surprisingly, glial cell specification was not affected upon Sox9 ablation. Our findings suggest Sox9 may mediate the neurogenic-to-gliogenic fate switch in mouse cerebellum by modulating the termination of neurogenesis, and therefore indicate a functional discrepancy of Sox9 between the development of cerebellum and other major neural tissues.
Sigma-Aldrich
Anti-Sox2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Brain lipid binding protein Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-LIM-1 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, (H+L) HRP conjugate, 1 mg/mL, Chemicon®
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, Peroxidase Conjugated, 1 mg/mL (after reconstitution), Chemicon®
Sigma-Aldrich
Anti-Sox9 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Ki-67 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-PAX6 Antibody, from rabbit, purified by affinity chromatography