Merck
Metformin as adjunct antituberculosis therapy.
Science translational medicine (2014-11-21)
Amit Singhal, Liu Jie, Pavanish Kumar, Gan Suay Hong, Melvin Khee-Shing Leow, Bhairav Paleja, Liana Tsenova, Natalia Kurepina, Jinmiao Chen, Francesca Zolezzi, Barry Kreiswirth, Michael Poidinger, Cynthia Chee, Gilla Kaplan, Yee Tang Wang, Gennaro De Libero
ABSTRACT
The global burden of tuberculosis (TB) morbidity and mortality remains immense. A potential new approach to TB therapy is to augment protective host immune responses. We report that the antidiabetic drug metformin (MET) reduces the intracellular growth of Mycobacterium tuberculosis (Mtb) in an AMPK (adenosine monophosphate-activated protein kinase)-dependent manner. MET controls the growth of drug-resistant Mtb strains, increases production of mitochondrial reactive oxygen species, and facilitates phagosome-lysosome fusion. In Mtb-infected mice, use of MET ameliorated lung pathology, reduced chronic inflammation, and enhanced the specific immune response and the efficacy of conventional TB drugs. Moreover, in two separate human cohorts, MET treatment was associated with improved control of Mtb infection and decreased disease severity. Collectively, these data indicate that MET is a promising candidate host-adjunctive therapy for improving the effective treatment of TB.
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Supelco
Loperamide hydrochloride, VETRANAL®, analytical standard
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Loperamide hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Loperamide hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
(±)-Verapamil hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
1,1-Dimethylbiguanide hydrochloride, 97%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Loperamide hydrochloride
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
(±)-Verapamil hydrochloride, ≥99% (titration), powder
Sigma-Aldrich
Ethionamide
Sigma-Aldrich
(±)-Bay K8644, calcium channel agonist
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
FPL 64176, ≥98% (HPLC), powder
Sigma-Aldrich
2′,3′-Dideoxyadenosine, ≥97% (HPLC)
Sigma-Aldrich
Diethyl azodicarboxylate solution, purum, ~40% in toluene (H-NMR)
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Streptomycin sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O