Merck
  • Taurine chloramine-induced inactivation of cofilin protein through methionine oxidation.
Taurine chloramine-induced inactivation of cofilin protein through methionine oxidation.
Free radical biology & medicine (2014-07-25)
Shen Luo, Hiroshi Uehara, Emily Shacter
ABSTRACT
Cofilin regulates reorganization of actin filaments (F-actin) in eukaryotes. A recent finding has demonstrated that oxidation of cofilin by taurine chloramine (TnCl), a physiological oxidant derived from neutrophils, causes cofilin to translocate to the mitochondria inducing apoptosis (F. Klamt et al. Nat. Cell Biol.11:1241-1246; 2009). Here we investigated the effect of TnCl on biological activities of cofilin in vitro. Our data show that TnCl-induced oxidation of recombinant human cofilin-1 inhibits its F-actin-binding and depolymerization activities. Native cofilin contains four free Cys and three Met residues. Incubation of oxidized cofilin with DTT does not lead to its reactivation. A double Cys to Ala mutation on the two C-terminal Cys shows similar biological activities as the wild type, but does not prevent the TnCl-induced inactivation. In contrast, incubation of oxidized cofilin with methionine sulfoxide reductases results in its reactivation. Phosphorylation is known to inhibit cofilin activities. We found that Met oxidation also prevents phosphorylation of cofilin, which is reversed by incubating oxidized cofilin with methionine sulfoxide reductases. Interestingly, intact protein mass spectrometry of the oxidized mutant indicated one major oxidation product with an additional mass of 16 Da, consistent with oxidation of one specific Met residue. This residue was identified as Met-115 by peptide mapping and tandem mass spectrometry. It is adjacent to Lys-114, a known residue on globular-actin-binding site, implying that oxidation of Met-115 disrupts the globular-actin-binding site of cofilin, which causes TnCl-induced inactivation. The findings identify Met-115 as a redox switch on cofilin that regulates its biological activity.
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
SAFC
BIS-TRIS
SAFC
BIS-TRIS
Supelco
Taurine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Taurine, ≥98%, FG
SAFC
Taurine
Sigma-Aldrich
Taurine, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Taurine, ≥99%
Sigma-Aldrich
Taurine, suitable for cell culture, meets USP testing specifications
USP
Taurine, United States Pharmacopeia (USP) Reference Standard