Merck
  • Histamine H4 and H1 receptors contribute to postinflammatory visceral hypersensitivity.
Histamine H4 and H1 receptors contribute to postinflammatory visceral hypersensitivity.
Gut (2014-02-25)
Annemie Deiteren, Joris G De Man, Nathalie E Ruyssers, Tom G Moreels, Paul A Pelckmans, Benedicte Y De Winter
ABSTRACT
Substantial evidence implicates mast cells and their main constituent histamine in the pathogenesis of visceral hypersensitivity. We explored the specific contribution of histamine H4 (H4R) and H1 (H1R) receptors to visceral hypersensitivity in a postinflammatory rat model. Trinitrobenzenesulfonic acid (TNBS)-colitis was monitored individually by colonoscopy: first on day 3 to confirm the presence of colitis and then every 4 days, starting from day 10, to monitor convalescence and determine the exact timepoint of endoscopic healing in each rat. Experiments were performed 3 days after endoscopic resolution of colitis. Visceral sensitivity was assessed by quantifying visceromotor responses (VMRs) to colorectal distension. Colonic mast cell numbers, histamine release and H4R and H1R mRNA expression were quantified. JNJ7777120 (H4R antagonist) and/or levocetirizine (H1R antagonist) were administered 30 min prior to VMR assessment or histamine release assay. Postcolitis rats displayed a higher number of colonic mast cells, excessive histamine release and significantly enhanced VMRs. Heightened VMRs were dose-dependently reduced by JNJ7777120 and levocetirizine; combined administration of JNJ7777120 and levocetirizine potentiated the antinociceptive effect. In the colon, both H4R and H1R mRNA were present; in the dorsal root ganglia, only H1R mRNA was found. Only colonic H4R mRNA expression was increased in postcolitis rats. Excessive histamine release in postcolitis rats was attenuated by the highest dose of JNJ7777120. H4R and H1R antagonists dose-dependently reduce and even normalise postinflammatory visceral hypersensitivity via different underlying mechanisms but with a synergistic effect. Both receptor subtypes represent promising targets for the treatment of postinflammatory visceral hypersensitivity.
Sigma-Aldrich
Cetirizine dihydrochloride, ≥98% (HPLC)
Cetirizine for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
o-Dianisidine dihydrochloride, Suitable for use in glucose determination
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
o-Dianisidine dihydrochloride, ≥95%
Sigma-Aldrich
Histamine, ≥97.0%
Sigma-Aldrich
Human IL-2 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
USP
Cetrimonium bromide, United States Pharmacopeia (USP) Reference Standard
Cetirizine dihydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
o-Dianisidine dihydrochloride, tablet, 10 mg substrate per tablet
Supelco
Histamine, analytical standard
Sigma-Aldrich
Bovine IL2 / Interleukin-2 ELISA Kit, for serum, plasma and cell culture supernatants
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Cetirizin dihydrochloride, ≥98.0% (HPLC)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
Picrylsulfonic acid solution, 5 % (w/v) in H2O, BioReagent, suitable for determination of primary amines