Merck
  • Dlg5 regulates dendritic spine formation and synaptogenesis by controlling subcellular N-cadherin localization.
Dlg5 regulates dendritic spine formation and synaptogenesis by controlling subcellular N-cadherin localization.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2014-09-19)
Shih-Hsiu J Wang, Ivana Celic, Se-Young Choi, Martin Riccomagno, Qiang Wang, Lu O Sun, Sarah P Mitchell, Valera Vasioukhin, Richard L Huganir, Alex L Kolodkin
ABSTRACT
Most excitatory synapses in the mammalian brain are formed on dendritic spines, and spine density has a profound impact on synaptic transmission, integration, and plasticity. Membrane-associated guanylate kinase (MAGUK) proteins are intracellular scaffolding proteins with well established roles in synapse function. However, whether MAGUK proteins are required for the formation of dendritic spines in vivo is unclear. We isolated a novel disc large-5 (Dlg5) allele in mice, Dlg5(LP), which harbors a missense mutation in the DLG5 SH3 domain, greatly attenuating its ability to interact with the DLG5 GUK domain. We show here that DLG5 is a MAGUK protein that regulates spine formation, synaptogenesis, and synaptic transmission in cortical neurons. DLG5 regulates synaptogenesis by enhancing the cell surface localization of N-cadherin, revealing a key molecular mechanism for regulating the subcellular localization of this cell adhesion molecule during synaptogenesis.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-V5 antibody, Mouse monoclonal, clone V5-10, purified from hybridoma cell culture
Sigma-Aldrich
Anti-POU3F2 antibody produced in rabbit
Sigma-Aldrich
Monoclonal Anti-β-Catenin antibody produced in mouse, clone 15B8, ascites fluid
Sigma-Aldrich
Anti-SLC32A1 (N-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-PSD95 Antibody, clone K28/43, clone K28/43, from mouse
Sigma-Aldrich
Anti-Vesicular Glutamate Transporter 1 Antibody, serum, Chemicon®