Merck
  • Using high-performance ¹H NMR (HP-qNMR®) for the certification of organic reference materials under accreditation guidelines--describing the overall process with focus on homogeneity and stability assessment.
Using high-performance ¹H NMR (HP-qNMR®) for the certification of organic reference materials under accreditation guidelines--describing the overall process with focus on homogeneity and stability assessment.
Journal of pharmaceutical and biomedical analysis (2013-11-05)
Michael Weber, Christine Hellriegel, Alexander Rueck, Juerg Wuethrich, Peter Jenks
ABSTRACT
Quantitative NMR spectroscopy (qNMR) is gaining interest across both analytical and industrial research applications and has become an essential tool for the content assignment and quantitative determination of impurities. The key benefits of using qNMR as measurement method for the purity determination of organic molecules are discussed, with emphasis on the ability to establish traceability to "The International System of Units" (SI). The work describes a routine certification procedure from the point of view of a commercial producer of certified reference materials (CRM) under ISO/IEC 17025 and ISO Guide 34 accreditation, that resulted in a set of essential references for (1)H qNMR measurements, and the relevant application data for these substances are given. The overall process includes specific selection criteria, pre-tests, experimental conditions, homogeneity and stability studies. The advantages of an accelerated stability study over the classical stability-test design are shown with respect to shelf-life determination and shipping conditions.
Product Description
Supelco
1,2,4,5-Tetramethylbenzene, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Dimethyl sulfone, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Dimethyl terephthalate, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Ethyl 4-(dimethylamino)benzoate, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Ethylene carbonate, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Calcium formate, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Potassium phthalate monobasic, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Maleic acid, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
1,2,4,5-Tetrachloro-3-nitrobenzene, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Benzoic acid, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Potassium phthalate monobasic, puriss. p.a., ≥99.5% (T)
Supelco
L-Glutamic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Dimethylmalonic acid, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Thymol, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Phosphonoacetic acid, TraceCERT®, 31P-qNMR Standard, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Methyl 3,5-dinitrobenzoate, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Triphenyl phosphate, TraceCERT®, 31P-qNMR Standard, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland