Merck
  • Heparin-binding growth-associated molecule contains two heparin-binding beta -sheet domains that are homologous to the thrombospondin type I repeat.
Heparin-binding growth-associated molecule contains two heparin-binding beta -sheet domains that are homologous to the thrombospondin type I repeat.
The Journal of biological chemistry (2000-05-02)
I Kilpelainen, M Kaksonen, T Kinnunen, H Avikainen, M Fath, R J Linhardt, E Raulo, H Rauvala
ABSTRACT
Heparin-binding growth-associated molecule (HB-GAM) is an extracellular matrix-associated protein implicated in the development and plasticity of neuronal connections of brain. Binding to cell surface heparan sulfate is indispensable for the biological activity of HB-GAM. In the present paper we have studied the structure of recombinant HB-GAM using heteronuclear NMR. These studies show that HB-GAM contains two beta-sheet domains connected by a flexible linker. Both of these domains contain three antiparallel beta-strands. In addition to this domain structure, HB-GAM contains the N- and C-terminal lysine-rich sequences that lack a detectable structure and appear to form random coils. Studies using CD and NMR spectroscopy suggest that HB-GAM undergoes a conformational change upon binding to heparin, and that the binding occurs primarily to the beta-sheet domains of the protein. Search of sequence data bases shows that the beta-sheet domains of HB-GAM are homologous to the thrombospondin type I repeat (TSR). Sequence comparisions show that the beta-sheet structures found previously in midkine, a protein homologous with HB-GAM, also correspond to the TSR motif. We suggest that the TSR sequence motif found in various extracellular proteins defines a beta-sheet structure similar to that found in HB-GAM and midkine. In addition to the apparent structural similarity, a similarity in biological functions is suggested by the occurrence of the TSR sequence motif in a wide variety of proteins that mediate cell-to-extracellular matrix and cell-to-cell interactions, in which the TSR domain mediates specific cell surface binding.
Sigma-Aldrich
ISOGRO®-15N Powder -Growth Medium, 98 atom % 15N
Sigma-Aldrich
ISOGRO®-D Powder -Growth Medium, 97-99 atom % D
Sigma-Aldrich
ISOGRO®-15N,D Powder -Growth Medium, 98 atom % 15N, 97 atom % D
Sigma-Aldrich
ISOGRO®-13C,15N,D Powder -Growth Medium, 98 atom % 15N, 97-99 atom % D, 99 atom % 13C
Sigma-Aldrich
ISOGRO®-13C,15N Powder -Growth Medium, 98 atom % 15N, 99 atom % 13C
Sigma-Aldrich
ISOGRO®-13C Powder -Growth Medium, 99 atom % 13C