Merck
  • Acidification of non-medicated and oxytetracycline-medicated cattle manures during anaerobic digestion.
Acidification of non-medicated and oxytetracycline-medicated cattle manures during anaerobic digestion.
Environmental technology (2014-08-26)
Cağri Akyol, Orhan Ince, Gökhan Türker, Bahar Ince
ABSTRACT
Possible adverse effects of a commonly used veterinary antibiotic, oxytetracycline (OTC), on acidogenic phase of anaerobic digestion of cattle manure along with optimum operating conditions were investigated. A standard veterinary practice of 50 ml OTC solution (20 mg/kg cattle weight) was injected into the muscles of cattle and then manure samples were collected for 5 days following the injection. The 5-day samples were equally mixed and used throughout digestion experiments. Preliminary batch tests were conducted to obtain the optimum pH range and observe volatile fatty acids (VFAs) production. In this regard, different sets of batch digesters were operated at pH ranging from 5.2 +/- 0.1 to 5.8 +/- 0.1 at mesophilic conditions with total solids content of 6.0 +/- 0.2%. The pH of 5.5 +/- 0.1 was found to be the optimum value for acidification for both non-medicated and OTC-medicated conditions. Under predetermined conditions, maximum total VFA (VFAtot) of 830 +/- 3 mg (as acetic acid)/L was produced and maximum acidification rate was evaluated as 11% for OTC-medicated cattle manure, whereas they were 900 +/- 6 mg (as acetic acid)/L and 12% for non-medicated manure. Digestion studies were further continued in a semi-continuous mode at pH 5.5 +/- 0.1 and SRT/HRT of 5 days. VFAtot concentrations and maximum acidification rate increased up to 2181 +/- 19 mg (as acetic acid)/L and 29% for non-medicated cattle manure. For OTC-medicated cattle manure, lower acidification rate of 18% was observed.
Product Description
Millipore
Oxytetra Selective Supplement, suitable for microbiology
Supelco
Oxytetracycline hydrochloride, VETRANAL®, analytical standard
USP
Oxytetracycline, United States Pharmacopeia (USP) Reference Standard
USP
Oxytetracycline hydrochloride, United States Pharmacopeia (USP) Reference Standard
Supelco
Oxytetracycline Hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Oxytetracycline hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Oxytetracycline hydrochloride
Sigma-Aldrich
Oxytetracycline dihydrate, Assay (anhydrous basis) 95.0%-102.0%