Merck
Single-walled carbon nanotubes used as stationary phase in GC.
Analytical chemistry (2006-09-15)
Li-Ming Yuan, Chao-Xing Ren, Li Li, Ping Ai, Zhi-Hong Yan, Min Zi, Zheng-Yu Li
ABSTRACT
Single-walled carbon nanotubes (SWNTs) have high surface area, high adsorption ability, and nanoscale interactions. In this study, capillary columns including SWNTs, ionic liquid (IL), and IL + SWNTs for GC were prepared. The separation results showed that SWNTs possessed a wide selectivity toward alkanes, alcohols, aromatic compounds, and ketones, and a SWNT capillary column was a very useful GC column for the separation of gas samples. Coating the IL stationary phase on the SWNT capillary column, the SWNTs were able to improve chromatographic characteristic of ionic liquid. Comparing the IL coated on three graphite carbon black capillary columns, which were prepared by dynamic coating, static coating, and chemical bonding the Carbopack C with on SWNTs capillary column, the capacity factors were much higher on the SWNT column. The SEM showed that SWNTs could be bonded to the inner surface of capillary tubing, and most of them were linked end-to-end to form a layer of network structure of skeletons resulting in a high surface area, which increased the interactions between stationary phase and analytes. This is the first single-wall carbon nanotubes bonded to the fused-silica capillary tubing. In the first approach, SWNTs assist ionic liquid with enhanced chromatographic characteristic in GC. This work indicates that SWNTs make it possible to extend the application range on the newly prepared chromatographic stationary phases for GC.
Product Description
Supelco
Carbopack Adsorbent, matrix Carbopack B, 60-80 mesh, bottle of 10 g
Supelco
Carbopack X, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
Carbopack Adsorbent, matrix Carbopack C, 80-100 mesh, bottle of 10 g
Supelco
Carbopack B, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
Carbopack Adsorbent, matrix Carbopack X, 40-60 mesh, bottle of 10 g
Supelco
Carbopack Adsorbent, matrix Carbopack X, 120-400 mesh, bottle of 50 g
Supelco
Carbopack Adsorbent, matrix Carbopack C, 60-80 mesh, bottle of 10 g
Supelco
Carbopack B, stainless steel TD tube, O.D. × L 1/4 × 3 1/2, unconditioned, pkg of 10 ea
Supelco
Carbopack B, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., unconditioned, pkg of 10 ea, matrix Carbopack
Supelco
Carbopack B, O.D. × L 1/4 in. × 3 1/2 in., glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
Supelco
Carbopack Adsorbent, matrix Carbopack X, 60-80 mesh, bottle of 10 g
Supelco
FLM Carbopack X Deactivated Stainless Steel TD Tube, preconditioned, pkg of 10 tubes, O.D. × L 1/4 in. (6.35 mm) × 3 1/2 in. (89 mm)