Merck
  • Clinical review of once-daily beclomethasone dipropionate for seasonal allergic rhinitis.
Clinical review of once-daily beclomethasone dipropionate for seasonal allergic rhinitis.
Clinical therapeutics (1996-09-01)
P Chervinsky
ABSTRACT
A new double-strength (84 micrograms/spray) formulation of beclomethasone dipropionate (BDP-ds) as an aqueous suspension has been introduced to control symptoms of allergic rhinitis with once-daily intranasal dosing. This paper reviews the results of three clinical reports which show that BDP-ds given once a day is more effective than placebo and as effective as regular-strength beclomethasone dipropionate given twice daily in reducing the nasal symptoms of seasonal allergic rhinitis. Symptom improvement was seen within 2 days of treatment initiation and was maintained for the subsequent 4 weeks. BDP-ds was found to be as safe and well tolerated as placebo. Adverse effects, which were generally mild, included headache, nasal burning/irritation, epistaxis, coughing, and pharyngitis. BDP-ds is safe for children as young as 6 years of age, and its once-daily dosing schedule may improve patient compliance.
Product Description
Beclometasone dipropionate for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Beclomethasone dipropionate, United States Pharmacopeia (USP) Reference Standard
Supelco
Beclomethasone dipropionate, analytical standard, for drug analysis
Beclometasone dipropionate for peak identification, European Pharmacopoeia (EP) Reference Standard
Beclomethasone dipropionate, anhydrous, European Pharmacopoeia (EP) Reference Standard